Imprimer

Anti-TLR3 Mouse Monoclonal Antibody [clone: 303CT19.9.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031297EA 535 EUR
USBI031297
Anti-TLR3 Mouse Monoclonal Antibody [clone: 303CT19.9.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: TLR3 (toll-like receptor 3)
Clonality: Monoclonal
Clone: 303CT19.9.2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR