Imprimer

Anti-TRAF2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 214CT16.3.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031306-HRPEA 720 EUR
USBI031306-HRP
Anti-TRAF2 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 214CT16.3.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-1000; Immunofluorescence: 1:10-50

Type: Primary
Antigen: TRAF2 (TNF receptor-associated factor 2)
Clonality: Monoclonal
Clone: 214CT16.3.4
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR