Imprimer

Anti-TSC1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 357CT4.4.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031310-FITCEA 852 EUR
USBI031310-FITC
Anti-TSC1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 357CT4.4.2]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: TSC1 (tuberous sclerosis 1)
Clonality: Monoclonal
Clone: 357CT4.4.2
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR