Imprimer

Anti-TSC2 Mouse Monoclonal Antibody [clone: 249CT19.1.3]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031312EA 527 EUR
USBI031312
Anti-TSC2 Mouse Monoclonal Antibody [clone: 249CT19.1.3]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100; Immunofluorescence: 1:10-50

Type: Primary
Antigen: TSC2 (tuberous sclerosis 2)
Clonality: Monoclonal
Clone: 249CT19.1.3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR