Imprimer

Anti-VEGFC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 30AT1174.77]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031327-PEEA 720 EUR
USBI031327-PE
Anti-VEGFC Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 30AT1174.77]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze! Light sensitive

Type: Primary
Antigen: VEGFC (vascular endothelial growth factor C)
Clonality: Monoclonal
Clone: 30AT1174.77
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR