Imprimer

Anti-FIGF Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 28AT743.288.48]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031328-FITCEA 852 EUR
USBI031328-FITC
Anti-FIGF Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 28AT743.288.48]
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: FIGF (c-fos induced growth factor (vascular endothelial growth factor D))
Clonality: Monoclonal
Clone: 28AT743.288.48
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR