Imprimer

Anti-FLT4 Mouse Monoclonal Antibody [clone: 44CT92.4.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031332EA 504 EUR
USBI031332
Anti-FLT4 Mouse Monoclonal Antibody [clone: 44CT92.4.4]
Anticorps
Recommended dilution: Western Blot: 1:2000; Immunochemistry: 1:10-1:50

Type: Primary
Antigen: FLT4 (fms-related tyrosine kinase 4)
Clonality: Monoclonal
Clone: 44CT92.4.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR