Imprimer

Anti-YEATS4 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 204CT4.7.2]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031339-APEA 711 EUR
USBI031339-AP
Anti-YEATS4 Mouse Monoclonal Antibody (AP (Alkaline Phosphatase)) [clone: 204CT4.7.2]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-250

Type: Primary
Antigen: YEATS4 (YEATS domain containing 4)
Clonality: Monoclonal
Clone: 204CT4.7.2
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR