Imprimer

Anti-ZWINT Mouse Monoclonal Antibody [clone: 422CT4.6.4]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031345EA 527 EUR
USBI031345
Anti-ZWINT Mouse Monoclonal Antibody [clone: 422CT4.6.4]
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:500-4000

Type: Primary
Antigen: ZWINT (ZW10 interacting kinetochore protein)
Clonality: Monoclonal
Clone: 422CT4.6.4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR