Imprimer

Anti-Cherry Chicken Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031351EA 401 EUR
USBI031351
Anti-Cherry Chicken Polyclonal Antibody
Anticorps
Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:2000

Type: Tag
Antigen: Cherry
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR