Imprimer

Anti-HSPB1 Mouse Monoclonal Antibody [clone: 13B825]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031373EA 432 EUR
USBI031373
Anti-HSPB1 Mouse Monoclonal Antibody [clone: 13B825]
Anticorps
Recommended dilution: Immunofluorescence: 1:1000; Western Blot: 1:10000

Type: Primary
Antigen: HSPB1 (heat shock 27kDa protein 1)
Clonality: Monoclonal
Clone: 13B825
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Human, Mouse, Pig, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR