Imprimer

Anti-Muscleblind Mouse Monoclonal Antibody [clone: 13B834]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031391EA 401 EUR
USBI031391
Anti-Muscleblind Mouse Monoclonal Antibody [clone: 13B834]
Anticorps
Recommended dilution: Immunochemistry: 1:1000; Immunofluorescence: 1:50-1:250; Western Blot: 1:1000-1:5000

Type: Primary
Antigen: Muscleblind
Clonality: Monoclonal
Clone: 13B834
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human, Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR