Imprimer

Anti-Neurofilament L, 68kD Chicken Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031406EA 432 EUR
USBI031406
Anti-Neurofilament L, 68kD Chicken Polyclonal Antibody
Anticorps
Recommended dilution: Immunofluorescence: 1:5000; Western Blot: 1:10000

Type: Primary
Antigen: Neurofilament L, 68kD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse, Rat
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR