Imprimer

Anti-Nop5p Mouse Monoclonal Antibody [clone: 13B846]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031412EA 401 EUR
USBI031412
Anti-Nop5p Mouse Monoclonal Antibody [clone: 13B846]
Anticorps
Recommended dilution: Immunofluorescence: 1:1000-1:10000; Western Blot: 1:10000

Type: Primary
Antigen: Nop5p
Clonality: Monoclonal
Clone: 13B846
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Yeast
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR