Imprimer

Anti-Synuclein, alpha Mouse Monoclonal Antibody [clone: 13B860]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031430EA 432 EUR
USBI031430
Anti-Synuclein, alpha Mouse Monoclonal Antibody [clone: 13B860]
Anticorps
Type: Primary
Antigen: Synuclein, alpha
Clonality: Monoclonal
Clone: 13B860
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rodent
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR