Imprimer

Anti-ADAMTS10 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: US Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031611-BIOTINEA 702 EUR
USBI031611-BIOTIN
Anti-ADAMTS10 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ADAMTS10 (ADAM metallopeptidase with thrombospondin type 1 motif, 10)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR