Imprimer

Anti-AFF1 Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031671EA 518 EUR
USBI031671
Anti-AFF1 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000

Type: Primary
Antigen: AFF1 (AF4/FMR2 family, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR