Imprimer

Anti-ALOXE3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: US Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI031815-HRPEA 711 EUR
USBI031815-HRP
Anti-ALOXE3 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: ALOXE3 (arachidonate lipoxygenase 3)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR