Imprimer

Anti-SMAD2 Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042020EA 421 EUR
USBI042020
Anti-SMAD2 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Dot blot 1:500; Immunofluorescence: 1:10-50

Type: Primary
Antigen: SMAD2 (SMAD family member 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR