Imprimer

Anti-SMAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042030-FITCEA 619 EUR
USBI042030-FITC
Anti-SMAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SMAP1 (small ArfGAP 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR