Imprimer

Anti-SMCP Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042040-PEEA 619 EUR
USBI042040-PE
Anti-SMCP Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: SMCP (sperm mitochondria-associated cysteine-rich protein)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR