Imprimer

Anti-SNRK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042093-APEA 852 EUR
USBI042093-AP
Anti-SNRK Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorps
Type: Primary
Antigen: SNRK (SNF related kinase)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR