Imprimer

Anti-SNRNP40 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042096-BIOTINEA 619 EUR
USBI042096-BIOTIN
Anti-SNRNP40 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: Western Blot: 1:100-1:500; Immunochemistry: 1:50-1:100

Type: Primary
Antigen: SNRNP40 (small nuclear ribonucleoprotein 40kDa (U5))
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR