Imprimer

Anti-SNTB1 Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042104EA 495 EUR
USBI042104
Anti-SNTB1 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SNTB1 (syntrophin, beta 1 (dystrophin-associated protein A1, 59kDa, basic component 1))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR