Imprimer

Anti-SOST Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042148-HRPEA 619 EUR
USBI042148-HRP
Anti-SOST Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunochemistry: 1:10-50

Type: Primary
Antigen: SOST (sclerostin)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR