Imprimer

Anti-SOX1 Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042150EA 495 EUR
USBI042150
Anti-SOX1 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:50-100; Immunofluorescence: 1:10-50; Immunochemistry: 1:10-50

Type: Primary
Antigen: SOX1 (SRY (sex determining region Y)-box 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR