Imprimer

Anti-SOX2 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042166-BIOTINEA 852 EUR
USBI042166-BIOTIN
Anti-SOX2 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Immunofluorescence: 1:100; Dot blot 1:100-500

Type: Primary
Antigen: SOX2 (SRY (sex determining region Y)-box 2)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR