Imprimer

Anti-SP140 Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042182EA 495 EUR
USBI042182
Anti-SP140 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: Western Blot: 1:1000; Immunochemistry: 1:10-1:50; Immunofluorescence: 1:10-1:50

Type: Primary
Antigen: SP140 (SP140 nuclear body protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR