Imprimer

Anti-SPRED2 Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042262EA 495 EUR
USBI042262
Anti-SPRED2 Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: SPRED2 (sprouty-related, EVH1 domain containing 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR