Imprimer

Anti-SRRD Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042316-HRPEA 619 EUR
USBI042316-HRP
Anti-SRRD Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500

Type: Primary
Antigen: SRRD (SRR1 domain containing)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR