Imprimer

Anti-STAMBP Rabbit Polyclonal Antibody (Biotin)

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042367-BIOTINEA 619 EUR
USBI042367-BIOTIN
Anti-STAMBP Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50

Type: Primary
Antigen: STAMBP (STAM binding protein)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR