Imprimer

Anti-Staphylococcus aureus, delta Hemolysin Rabbit Polyclonal Antibody (Biotin)

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042370-BIOTINEA 619 EUR
USBI042370-BIOTIN
Anti-Staphylococcus aureus, delta Hemolysin Rabbit Polyclonal Antibody (Biotin)
Anticorps
Recommended dilution: Western Blot: 1:1000

Type: Primary
Antigen: Staphylococcus aureus, delta Hemolysin
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR