Imprimer

Anti-Staphylococcus aureus, delta Hemolysin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042370-FITCEA 619 EUR
USBI042370-FITC
Anti-Staphylococcus aureus, delta Hemolysin Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Recommended dilution: Western Blot: 1:1000
Do not freeze!

Type: Primary
Antigen: Staphylococcus aureus, delta Hemolysin
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR