Imprimer

Anti-STAU1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042386-APCEA 619 EUR
USBI042386-APC
Anti-STAU1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: STAU1 (staufen double-stranded RNA binding protein 1)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR