Imprimer

Anti-STK32A Rabbit Polyclonal Antibody (APC (Allophycocyanin))

Fournisseur: US Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042404-APCEA 619 EUR
USBI042404-APC
Anti-STK32A Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:50-100
Do not freeze!

Type: Primary
Antigen: STK32A (serine/threonine kinase 32A)
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR