Imprimer
Anti-STX19 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Fournisseur: US Biological
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
USBI042434-FITCEA 852 EUR
USBI042434-FITC
Anti-STX19 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorps
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500; Immunochemistry: 1:10-50
Do not freeze!
Type: Primary
Antigen: STX19 (syntaxin 19)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope: C-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human