Imprimer

Anti-TAS1R3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI042607-APEA 619 EUR
USBI042607-AP
Anti-TAS1R3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anticorps
Recommended dilution: ELISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TAS1R3 (taste receptor, type 1, member 3)
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR