Imprimer

Anti-FGD3 Mouse Monoclonal Antibody [clone: 4E6]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBI126759EA 522 EUR
USBI126759
Anti-FGD3 Mouse Monoclonal Antibody [clone: 4E6]
Anticorps
Type: Primary
Antigen: FGD3 (FYVE, RhoGEF and PH domain containing 3)
Clonality: monoclonal
Clone: 4000000
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR