Imprimer
Anti-SPHK1 Mouse Monoclonal Antibody [clone: 1D6]
Fournisseur: United States Biological
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
USBI133768EA 522 EUR
USBI133768
Anti-SPHK1 Mouse Monoclonal Antibody [clone: 1D6]
Anticorps
Type: Primary
Antigen: SPHK1 (sphingosine kinase 1)
Clonality: monoclonal
Clone: 1D6
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human