Imprimer

Anti-Brain Derived Neurotrophic Factor Mouse monoclonal antibody unconjugated [clone: 8J359]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIB2700-02LEA 716 EUR
USBIB2700-02L
Anti-Brain Derived Neurotrophic Factor Mouse monoclonal antibody unconjugated [clone: 8J359]
Anticorps
Recommended dilution: Western Blot: 1:100-1:2000

Type: Primary
Antigen: Brain Derived Neurotrophic Factor
Clonality: monoclonal
Clone: 8J359
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR