Imprimer

Anti-FCGR3A Mouse monoclonal antibody [clone: MEM-154]

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIC2267-62JEA 596 EUR
USBIC2267-62J
Anti-FCGR3A Mouse monoclonal antibody [clone: MEM-154]
Anticorps
Type: Primary
Antigen: FCGR3A (Fc fragment of IgG, low affinity IIIa, receptor (CD16a))
Clonality: Monoclonal
Clone: MEM-154
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR