Imprimer

Anti-RB1 Rabbit Polyclonal Antibody (Biotin)

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIR1660-05K3BIOTEA 852 EUR
USBIR1660-05K3BIOT
Anti-RB1 Rabbit Polyclonal Antibody (Biotin)
Anticorps
Type: Primary
Antigen: RB1 (retinoblastoma 1)
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR