Imprimer

Anti-Tenascin Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIT2550-23EA 729 EUR
USBIT2550-23
Anti-Tenascin Rabbit Polyclonal Antibody
Anticorps
Type: Primary
Antigen: Tenascin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Zebrafish
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR