Imprimer

Anti-TFRC Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIT8199-23AEA 462 EUR
USBIT8199-23A
Anti-TFRC Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: Western Blot: 1:500-1:1000; Immunochemistry: 1:50-1:200; ELISA: 1:5000-1:20000

Type: Primary
Antigen: TFRC (transferrin receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR