Imprimer

Anti-Tfr-2 Rabbit polyclonal antibody

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIT8199-49EA 319 EUR
USBIT8199-49
Anti-Tfr-2 Rabbit polyclonal antibody
Type: Primary
Antigen: Tfr-2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR