Imprimer

Anti-Transferrin-Transferrin Receptor Complex Mouse monoclonal antibody [clone: 5E820]

Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIT8199-72EA 548 EUR
USBIT8199-72
Anti-Transferrin-Transferrin Receptor Complex Mouse monoclonal antibody [clone: 5E820]
Type: Primary
Antigen: Transferrin-Transferrin Receptor Complex
Clonality: monoclonal
Clone: 5E820
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR