Imprimer
Anti-TGFA Rabbit Polyclonal Antibody
Fournisseur: United States Biological
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
USBIT8250-02B-20EA 337 EUR
USBIT8250-02B-20
Anti-TGFA Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: TGFA (transforming growth factor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant