Imprimer

Anti-TGFA Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIT8250-04EEA 592 EUR
USBIT8250-04E
Anti-TGFA Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: Western Blot: 1:500

Type: Primary
Antigen: TGFA (transforming growth factor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR