Imprimer

Anti-TGFA Rabbit Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIT8250-06DEA 530 EUR
USBIT8250-06D
Anti-TGFA Rabbit Polyclonal Antibody
Anticorps
Recommended dilution: Western Blot: 1:3000; Immunochemistry: 1:1000-1:5000

Type: Primary
Antigen: TGFA (transforming growth factor, alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR