Imprimer

Anti-VIM Chicken Polyclonal Antibody

Fournisseur: United States Biological
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

USBIV2122-02PEA 796 EUR
USBIV2122-02P
Anti-VIM Chicken Polyclonal Antibody
Anticorps
Recommended dilution: Immunochemistry: 1:1000-1:2000; Western Blot: 1:2000-1:5000

Type: Primary
Antigen: VIM (vimentin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR